AOB640097603 AOBChem

5-Chloro-2-fluoro-4-hydroxybenzaldehyde

  • MDL number MFCD22066584
  • Cas Number : 838856-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96.00
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-chloro-2-fluoro-4-hydroxybenzaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/31170.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart