AOB640097744 AOBChem

2,6-Difluoro-4-isopropyloxybenzaldehyde

  • MDL number MFCD30344672
  • Cas Number : 1781138-46-2
  • Molecular Weight : 200.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-6-difluoro-4-isopropyloxybenzaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/31749.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart