AOB640097817 AOBChem

Methyl 4-chloro-2,3-difluorobenzoate

  • MDL number MFCD14698003
  • Cas Number : 1214334-98-1
  • Molecular Weight : 206.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/methyl-4-chloro-2-3-difluorobenzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32175.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart