AOB640097821 AOBChem

4-Chloro-2-fluoro-3-methoxybenzoic acid

  • MDL number MFCD19687204
  • Cas Number : 1169870-80-7
  • Molecular Weight : 204.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-chloro-2-fluoro-3-methoxybenzoic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32191.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart