AOB640097878 AOBChem

5-Bromo-2-fluoro-4-methylphenol

  • MDL number MFCD12405415
  • Cas Number : 1111096-04-8
  • Molecular Weight : 205.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/5-bromo-2-fluoro-4-methylphenol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32402.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart