AOB945511687 AOBChem

5-Chloro-4-hydroxy-3-(trifluoromethyl)phenylboronic acid pinacol ester

  • MDL number MFCD30723165
  • Cas Number : 2121515-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-chloro-4-hydroxy-3-trifluoromethyl-phenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/33027.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart