AOB945511742 AOBChem

4-Chloro-2,5-difluorobenzoic acid ethyl ester

  • MDL number MFCD12025299
  • Cas Number : 879093-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-chloro-2-5-difluorobenzoic-acid-ethyl-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/22507.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart