AOB945511806 AOBChem

Methyl 3-fluoro-5-methylbenzoate

  • MDL number MFCD06204167
  • Cas Number : 660416-38-6
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/methyl-3-fluoro-5-methylbenzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32127.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart