AOB945511928 AOBChem

4-Benzyloxy-3-chloro-5-fluorobenzeneboronic acid pinacol ester

  • MDL number MFCD30529839
  • Cas Number : 2121515-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-benzyloxy-3-chloro-5-fluorobenzeneboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32650.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart