AOB945511984 AOBChem

5-Ethoxy-2-fluoro-4-iodopyridine

  • MDL number MFCD28795550
  • Cas Number : 1805067-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-ethoxy-2-fluoro-4-iodopyridine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32786.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart