AOB945512010 AOBChem

3-Fluoro-4-methoxy-5-trifluoromethylphenylboronic acid pinacol ester

  • MDL number MFCD30529865
  • Cas Number : 2121513-13-9
  • Molecular Weight : 320.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-fluoro-4-methoxy-5-trifluoromethylphenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32865.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart