AOB945512025 AOBChem

5-Bromo-3-(pyrrolidine-1-carbonyl)phenylboronic acid

  • MDL number MFCD30529873
  • Cas Number : 2121514-36-9
  • Molecular Weight : 297.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/5-bromo-3-pyrrolidine-1-carbonyl-phenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32905.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart