AOB947289180 AOBChem

4-Fluoro-3-isopropoxybenzoic acid methyl ester

  • MDL number MFCD23115006
  • Cas Number : 869990-48-7
  • Molecular Weight : 212.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-fluoro-3-isopropoxybenzoic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/33246.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart