AOB979588440 AOBChem

3-Bromo-5-(9H-carbazol-9-yl)-phenylboronic acid pinacol ester

  • MDL number MFCD30834043
  • Cas Number : 2121512-10-3
  • Molecular Weight : 448.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-bromo-5-9h-carbazol-9-yl-phenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/33332.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart