AOB979588502 AOBChem

[4-(Benzyloxy)-3,5-dimethylphenyl]methanol

  • MDL number MFCD06202697
  • Cas Number : 773868-60-3
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-benzyloxy-3-5-dimethylphenyl-methanol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/33259.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart