AOB979588584 AOBChem

3-(N,N-Diethylaminocarbonyl)phenylboronic acid, pinacol ester

  • MDL number MFCD08458202
  • Cas Number : 325142-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-n-n-diethylaminocarbonyl-phenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/18903.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart