AOB979588586 AOBChem

Ethyl 2-fluoro-4-(trifluoromethyl)benzoate

  • MDL number MFCD06204411
  • Cas Number : 702673-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/ethyl-2-fluoro-4-trifluoromethyl-benzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/33504.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart