AOBH039CD368 AOBChem

1-Bromo-5-(4-chlorophenyl)-2-methylbenzene

  • MDL number MFCD28977194
  • Cas Number : 1033760-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/1-bromo-5-4-chlorophenyl-2-methylbenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/25718.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart