AOBH03B6AB56 AOBChem

4-Chloro-3-fluoro-2-methylphenylboronic acid

  • MDL number MFCD23712910
  • Cas Number : 2377606-88-5
  • Molecular Weight : 188.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-chloro-3-fluoro-2-methylphenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/31433.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart