AOBH03D4EF28 AOBChem

4-Methyl-pyridine-3-carbaldehyde

  • MDL number MFCD07437997
  • Cas Number : 51227-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-methyl-pyridine-3-carbaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/20251.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart