AOBH042EEA68 AOBChem

(4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

  • MDL number MFCD26402399
  • Cas Number : 443776-91-8
  • Molecular Weight : 264.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-methoxy-3-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-methanol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/16230.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart