AOBH042EEA93 AOBChem

(4-((4-Methylpiperidin-1-yl)methyl)phenyl)boronic acid

  • MDL number MFCD16198059
  • Cas Number : 1334399-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-4-methylpiperidin-1-yl-methyl-phenyl-boronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/32889.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart