AOBH11E4FCBC AOBChem

Ethyl 2,3-Dichloro-6-methoxybenzoate

  • MDL number MFCD31619484
  • Cas Number : 2179038-50-5
  • Molecular Weight : 249.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/ethyl-2-3-dichloro-6-methoxybenzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34373.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart