AOBH11E4FCC6 AOBChem

2-Fluoro-4-methyl-5-phenylbenzaldehyde

  • MDL number MFCD31619494
  • Cas Number : 2221812-03-7
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-fluoro-4-methyl-5-phenylbenzaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34483.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart