AOBH11E4FCD0 AOBChem

2-Amino-3,5-dichlorophenylboronic acid pinacol ester

  • MDL number MFCD31619508
  • Molecular Weight : 287.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-amino-3-5-dichlorophenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34596.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart