AOBH11E4FCD1 AOBChem

2,3-Difluoro-1-methoxy-4-(methylsulfanyl)benzene

  • MDL number MFCD31619510
  • Cas Number : 2221812-13-9
  • Molecular Weight : 190.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-3-difluoro-1-methoxy-4-methylsulfanyl-benzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34611.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart