AOBH11E4FCEA AOBChem

5-Bromo-3-chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester

  • Molecular Weight : 365.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-bromo-3-chloro-2-fluoro-4-methoxyphenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34811.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart