AOBH11E4FD42 AOBChem

2-(3,5-Dimethoxyphenyl)-1,3-dioxolane

  • MDL number MFCD06208952
  • Cas Number : 140464-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-3-5-dimethoxyphenyl-1-3-dioxolane.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/21857.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart