AOBH11E4FD45 AOBChem

2-Bromo-6-(methoxymethoxy)naphthalene

  • MDL number MFCD20927323
  • Cas Number : 111359-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-bromo-6-methoxymethoxy-naphthalene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/22019.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart