AOBH11E4FDC1 AOBChem

1-Bromo-4,5-difluoro-2-(phenylmethoxy)benzene

  • MDL number MFCD31619446
  • Cas Number : 1698908-86-9
  • Molecular Weight : 299.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-bromo-4-5-difluoro-2-phenylmethoxy-benzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/33480.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart