AOBH11E4FDC9 AOBChem

Methyl 4-(benzyloxy)-2,5-difluorobenzoate

  • MDL number MFCD28503326
  • Cas Number : 1416176-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/methyl-4-benzyloxy-2-5-difluorobenzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34015.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart