AOBH11E4FDF2 AOBChem

1-Bromo-2-chloro-4-(methoxymethoxy)-benzene

  • MDL number MFCD31619368
  • Cas Number : 1300743-20-7
  • Molecular Weight : 251.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/1-bromo-2-chloro-4-methoxymethoxy-benzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/15710.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart