AOBH11E4FDFC AOBChem

3-Bromo-4-chlorobenzaldehyde ethylene acetal

  • MDL number MFCD31619356
  • Cas Number : 2092689-22-8
  • Molecular Weight : 263.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/3-bromo-4-chlorobenzaldehyde-ethylene-acetal.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/11148.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart