AOBH11E4FE14 AOBChem

Ethyl 4-bromo-5-fluoro-2-methylbenzoate

  • MDL number MFCD28739714
  • Cas Number : 1807043-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/ethyl-4-bromo-5-fluoro-2-methylbenzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34495.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart