AOBH11E4FE94 AOBChem

1,3-Dimethyl-2-fluoro-5-iodobenzene

  • MDL number MFCD00120480
  • Cas Number : 1416549-07-9
  • Molecular Weight : 250.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/1-3-dimethyl-2-fluoro-5-iodobenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34774.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart