AOBH11E4FEA7 AOBChem

4-Fluoro-3-(morpholinomethyl)phenylboronic acid pinacol ester

  • MDL number MFCD23098827
  • Cas Number : 2377610-63-2
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-fluoro-3-morpholinomethyl-phenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/34814.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart