AOBH30376628 AOBChem

5-Methylpyridine-2-carboxylic acid methyl ester

  • MDL number MFCD11040648
  • Cas Number : 29681-38-7
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/5-methylpyridine-2-carboxylic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/19290.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart