AOBH3CD9F0E3 AOBChem

3-Fluoro-4-(1,3,2-dioxaborolan-2-yl)benzoyl chloride

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-fluoro-4-1-3-2-dioxaborolan-2-yl-benzoyl-chloride.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/16533.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart