AOBH3CD9F0F3 AOBChem

2-(3-Bromo-2-chloro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-3-bromo-2-chloro-5-methylphenyl-4-4-5-5-tetramethyl-1-3-2-dioxaborolane.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35433.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart