AOBH3CD9F0FF AOBChem

2-(5-Bromo-2,3-difluoro-4-ethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-5-bromo-2-3-difluoro-4-ethoxyphenyl-4-4-5-5-tetramethyl-1-3-2-dioxaborolane.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35610.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart