AOBH3CD9F12D AOBChem

5-(6-bromo-3-chloro-2-fluorophenyl)oxazole

  • Cas Number : 2364584-88-1
  • Molecular Weight : 276.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-6-bromo-3-chloro-2-fluorophenyl-oxazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35953.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart