AOBH3CD9F151 AOBChem

5-(3,5-dichloro-4-methylphenyl)oxazole

  • Cas Number : 2364585-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-3-5-dichloro-4-methylphenyl-oxazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/36013.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart