AOBH3CD9F154 AOBChem

Isopropyl 2-chloro-5-fluoro-3-(trifluoromethyl)benzoate

  • Cas Number : 2364585-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/isopropyl-2-chloro-5-fluoro-3-trifluoromethyl-benzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/36033.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart