AOBH3CD9F15D AOBChem

6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol

  • MDL number MFCD28740790
  • Cas Number : 1805580-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/6-bromo-2-fluoro-3-trifluoromethoxy-phenol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/33661.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart