AOBH3CD9F16E AOBChem

2-Bromo-6-fluoro-3-(trifluoromethyl)benzamide

  • Cas Number : 2091605-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-bromo-6-fluoro-3-trifluoromethyl-benzamide.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35458.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart