AOBH3CD9F190 AOBChem

3,6-Dimethoxy-4-pyridazinecarboxylic acid ethyl ester

  • MDL number MFCD29072142
  • Cas Number : 262353-17-3
  • Molecular Weight : 212.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-6-dimethoxy-4-pyridazinecarboxylic-acid-ethyl-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35596.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart