AOBH3CD9F19D AOBChem

Methyl 3-bromo-2-chloro-6-fluorobenzoate

  • MDL number MFCD28140551
  • Cas Number : 1784053-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/methyl-3-bromo-2-chloro-6-fluorobenzoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35612.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart