AOBH3CD9F1A1 AOBChem

3-(Benzyloxy)-5-chlorobenzaldehyde

  • MDL number MFCD16618949
  • Cas Number : 1228956-94-2
  • Molecular Weight : 246.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-benzyloxy-5-chlorobenzaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35619.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart