AOBH3CD9F1B1 AOBChem

5-[3-Bromo-5-(trifluoromethyl)phenyl]-oxazole

  • Cas Number : 1630747-25-9
  • Molecular Weight : 292.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-3-bromo-5-trifluoromethyl-phenyl-oxazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35710.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart