AOBH3CD9F1B8 AOBChem

5-(4-Chloro-3-ffluorophenyl)-1,3-oxazole

  • Cas Number : 2002638-39-1
  • Molecular Weight : 197.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-4-chloro-3-ffluorophenyl-1-3-oxazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35719.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart